1-Methyl-1H-pyrazole-4-sulfonyl chloride

Supplier: Matrix Scientific

033396-1G
101835-592EA 52.31 USD
101835-592
1-Methyl-1H-pyrazole-4-sulfonyl chloride
1-Methyl-1H-pyrazole-4-sulfonyl chloride

Hazard: Corrosive/Moisture Sensitive/Refrigerate

Formula: C₄H₅ClN₂O₂S
MW: 179.98 g/mol
Melting Pt: 46…48 °C
MDL Number: MFCD04968667
CAS Number: 288148-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR