2-(Allyloxy)-3-methoxybenzenecarbaldehyde

Supplier: Matrix Scientific

Synonyms: 3-Methoxy-2-(prop-2-en-1-yloxy)benzaldehyde

033376-500MG
101835-552EA 230 USD
101835-552
2-(Allyloxy)-3-methoxybenzenecarbaldehyde
2-(Allyloxy)-3-methoxybenzenecarbaldehyde

Hazard: Irritant

Formula: C₁₁H₁₂O₃
MW: 192.22 g/mol
MDL Number: MFCD03001271
CAS Number: 23343-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR