3-Chloro-4-(4-methylpiperazin-1-yl)phenylamine hydrochloride

Supplier: Matrix Scientific

033336-500MG
101835-472EA 89.7 USD
101835-472
3-Chloro-4-(4-methylpiperazin-1-yl)phenylamine hydrochloride
3-Chloro-4-(4-methylpiperazin-1-yl)phenylamine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₇Cl₂N₃
MDL Number: MFCD11506491
CAS Number: 1052538-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR