3,5-Dimethyl-1H-indole-2-carbaldehyde

Supplier: Matrix Scientific

033268-500MG
101835-340EA 230 USD
101835-340
3,5-Dimethyl-1H-indole-2-carbaldehyde
3,5-Dimethyl-1H-indole-2-carbaldehyde

Hazard: Irritant

Formula: C₁₁H₁₁NO
MDL Number: MFCD07643177
CAS Number: 1463-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR