1-Cyclopropylethylamine hydrochloride

Supplier: Matrix Scientific

Synonyms: 1-Cyclopropylethanamine hydrochloride

033263-500MG
101835-330EA 28.75 USD
101835-330
1-Cyclopropylethylamine hydrochloride
1-Cyclopropylethylamine hydrochloride

Hazard: Irritant

Formula: C₅H₁₂ClN
MW: 121.61 g/mol
MDL Number: MFCD06685893
CAS Number: 42390-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR