3-Methoxymethylpiperidine

Supplier: Matrix Scientific

033253-500MG
101835-310EA 287.5 USD
101835-310
3-Methoxymethylpiperidine
3-Methoxymethylpiperidine

Hazard: Irritant

Formula: C₇H₁₅NO
MDL Number: MFCD06248040
CAS Number: 116574-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR