4-(2-Aminothiazol-4-yl)benzoic acid

Supplier: Matrix Scientific

Synonyms: 4-(2-Amino-4-thiazolyl)benzoic Acid

033243-500MG
101835-292EA 230 USD
101835-292
4-(2-Aminothiazol-4-yl)benzoic acid
4-(2-Aminothiazol-4-yl)benzoic acid

Hazard: Irritant

Formula: C₁₀H₈N₂O₂S
MW: 220.25 g/mol
MDL Number: MFCD01460736
CAS Number: 216959-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR