4-Amino-5-pyridin-3-yl-4H-[1,2,4]triazole-3-thiol

Supplier: Matrix Scientific

033235-500MG
101835-276EA 230 USD
101835-276
4-Amino-5-pyridin-3-yl-4H-[1,2,4]triazole-3-thiol
4-Amino-5-pyridin-3-yl-4H-[1,2,4]triazole-3-thiol

Hazard: Irritant

Formula: C₇H₇N₅S
MDL Number: MFCD00834356
CAS Number: 78027-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR