2,4,5-Trimethoxybenzylamine

Supplier: Matrix Scientific

033222-500MG
101835-250EA 230 USD
101835-250
2,4,5-Trimethoxybenzylamine
2,4,5-Trimethoxybenzylamine

Hazard: Irritant

Formula: C₁₀H₁₅NO₃
MDL Number: MFCD03411011
CAS Number: 154584-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR