4-(4-Methylpiperazin-1-yl)benzoic acid

Supplier: Matrix Scientific

Synonyms: 1-(4-Carboxyphenyl)-4-methylpiperazine4-(4-Methylpiperazinyl)benzoic acid

033210-500MG
101835-226EA 141.54 USD
101835-226
4-(4-Methylpiperazin-1-yl)benzoic acid
4-(4-Methylpiperazin-1-yl)benzoic acid

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₂
MW: 220.2700 g/mol
MDL Number: MFCD02682063
CAS Number: 86620-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR