3,4-Dihydro-2H-benzo[b][1,4]dioxepine-7-sulfonyl chloride

Supplier: Matrix Scientific

033209-500MG
101835-224EA 149.5 USD
101835-224
3,4-Dihydro-2H-benzo[b][1,4]dioxepine-7-sulfonyl chloride
3,4-Dihydro-2H-benzo[b][1,4]dioxepine-7-sulfonyl chloride

Hazard: Irritant

Formula: C₉H₉ClO₄S
MW: 248.69 g/mol
MDL Number: MFCD02681959
CAS Number: 321309-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR