5-(2-Chlorophenyl)-2H-pyrazol-3-ylamine hydrochloride

Supplier: Matrix Scientific

033203-500MG
101835-212EA 89.7 USD
101835-212
5-(2-Chlorophenyl)-2H-pyrazol-3-ylamine hydrochloride
5-(2-Chlorophenyl)-2H-pyrazol-3-ylamine hydrochloride

Hazard: Irritant

Formula: C₉H₉Cl₂N₃
MDL Number: MFCD11506469
CAS Number: 1031791-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR