2-[2-(4-Methoxyphenyl)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
033192-500MG
101835-190EA
345
USD
101835-190
2-[2-(4-Methoxyphenyl)ethyl]piperidine hydrochloride
2-[2-(4-Methoxyphenyl)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₂ClNO |
MDL Number:
MFCD11506467 CAS Number: 1185294-00-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...