Cyclopentylhydrazine dihydrochloride

Supplier: Matrix Scientific

033176-500MG
101835-156EA 230 USD
101835-156
Cyclopentylhydrazine dihydrochloride
Cyclopentylhydrazine dihydrochloride

Hazard: Irritant

Formula: C₅H₁₄Cl₂N₂
MDL Number: MFCD06800488
CAS Number: 645372-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR