6-(1H-pyrazol-1-yl)nicotinic acid

Supplier: Matrix Scientific

Synonyms: 6-(1H-Pyrazol-1-yl)nicotinic acid

033160-500MG
101835-124EA 282.9 USD
101835-124
6-(1H-pyrazol-1-yl)nicotinic acid
6-(1H-pyrazol-1-yl)nicotinic acid

Hazard: Irritant

Formula: C₉H₇N₃O₂
MW: 189.174 g/mol
MDL Number: MFCD00140749
CAS Number: 253315-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR