5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulfonyl chloride

Supplier: Matrix Scientific

033154-500MG
101835-112EA 58.46 USD
101835-112
5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulfonyl chloride
5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulphonyl chloride

Hazard: Irritant

Formula: C₅H₆Cl₂N₂O₂S
MDL Number: MFCD00052536
CAS Number: 88398-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR