1-Ethyl-2-methyl-1H-indole-3-carbaldehyde

Supplier: Matrix Scientific

033139-500MG
101835-084EA 316.25 USD
101835-084
1-Ethyl-2-methyl-1H-indole-3-carbaldehyde
1-Ethyl-2-methyl-1H-indole-3-carbaldehyde

Hazard: Irritant

Formula: C₁₂H₁₃NO
MDL Number: MFCD03906275
CAS Number: 66727-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR