3-Chloro-6,7,8,9-tetrahydro-5H-carbazole-2-carboxylic acid

Supplier: Matrix Scientific

033114-500MG
101835-038EA 282.9 USD
101835-038
3-Chloro-6,7,8,9-tetrahydro-5H-carbazole-2-carboxylic acid
3-Chloro-6,7,8,9-tetrahydro-5H-carbazole-2-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₂ClNO₂
MDL Number: MFCD05625691
CAS Number: 676470-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR