5-(4-Fluorophenyl)furan-2-carboxylic acid

Supplier: Matrix Scientific

033082-500MG
101834-974EA 385.25 USD
101834-974
5-(4-Fluorophenyl)furan-2-carboxylic acid
5-(4-Fluorophenyl)furan-2-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₇FO₃
MDL Number: MFCD02104577
CAS Number: 73269-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR