6-(2-Bromoethyl)-1,3-dimethyl-1,6-dihydropyrrolo[3,4-d]pyrimidine-2,4-dione

Supplier: Matrix Scientific

033061-500MG
101834-936EA 230 USD
101834-936
6-(2-Bromoethyl)-1,3-dimethyl-1,6-dihydropyrrolo[3,4-d]pyrimidine-2,4-dione
6-(2-Bromoethyl)-1,3-dimethyl-1,6-dihydropyrrolo[3,4-d]pyrimidine-2,4-dione

Hazard: Irritant

Formula: C₁₀H₁₂BrN₃O₂
MDL Number: MFCD04011652
CAS Number: 841208-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR