(1-Benzyl-3,5-dimethyl-1H-pyrazol-4-yl)acetic acid

Supplier: Matrix Scientific

033046-500MG
101834-908EA 136.85 USD
101834-908
(1-Benzyl-3,5-dimethyl-1H-pyrazol-4-yl)acetic acid
(1-Benzyl-3,5-dimethyl-1H-pyrazol-4-yl)acetic acid

Hazard: Irritant

Formula: C₁₄H₁₆N₂O₂
MDL Number: MFCD06589791
CAS Number: 32701-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR