2-Methyl-4-morpholin-4-ylbenzaldehyde

Supplier: Matrix Scientific

033040-500MG
101834-902EA 230 USD
101834-902
2-Methyl-4-morpholin-4-ylbenzaldehyde
2-Methyl-4-morpholin-4-ylbenzaldehyde

Hazard: Irritant

Formula: C₁₂H₁₅NO₂
MDL Number: MFCD06589789
CAS Number: 736991-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR