2-Methoxypyrimidine-5-carbaldehyde

Supplier: Matrix Scientific

033021-500MG
101834-868EA 84.62 USD
101834-868
2-Methoxypyrimidine-5-carbaldehyde
2-Methoxypyrimidine-5-carbaldehyde

Hazard: Irritant

Formula: C₆H₆N₂O₂
MW: 138.1200 g/mol
MDL Number: MFCD03446732
CAS Number: 90905-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR