5-Chloro-1,3-dimethyl-1H-indole-2-carboxylic acid

Supplier: Matrix Scientific

033018-500MG
101834-862EA 230 USD
101834-862
5-Chloro-1,3-dimethyl-1H-indole-2-carboxylic acid
5-Chloro-1,3-dimethyl-1H-indole-2-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₀ClNO₂
MDL Number: MFCD03011751
CAS Number: 152088-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR