3-Methyl-7-nitro-1H-indole-2-carboxylic acid

Supplier: Matrix Scientific

033015-500MG
101834-856EA 184 USD
101834-856
3-Methyl-7-nitro-1H-indole-2-carboxylic acid
3-Methyl-7-nitro-1H-indole-2-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₈N₂O₄
MDL Number: MFCD03011739
CAS Number: 115058-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR