3-Chloro-6-(1-imidazolyl)pyridazine

Supplier: Matrix Scientific

033008-500MG
101834-842EA 235.38 USD
101834-842
3-Chloro-6-(1-imidazolyl)pyridazine
3-Chloro-6-(1-imidazolyl)pyridazine

Hazard: Irritant

Formula: C₇H₅ClN₄
MDL Number: MFCD02323226
CAS Number: 71037-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR