1-Isopropyl-2-piperidin-3-yl-1H-benzoimidazole dihydrochloride

Supplier: Matrix Scientific

033003-500MG
101834-832EA 152.95 USD
101834-832
1-Isopropyl-2-piperidin-3-yl-1H-benzoimidazole dihydrochloride
1-Isopropyl-2-piperidin-3-yl-1H-benzoimidazole dihydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃Cl₂N₃
MDL Number: MFCD11506440
CAS Number: 1185300-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR