1-((4-Chlorophenyl)sulfonyl)-4-pyridinecarboxylic acid

Supplier: Matrix Scientific

Synonyms: 1-((4-Chlorophenyl)sulfonyl)piperidine-4-carboxylic acid

032997-500MG
101834-820EA 148.35 USD
101834-820
1-((4-Chlorophenyl)sulfonyl)-4-pyridinecarboxylic acid
1-((4-Chlorophenyl)sulphonyl)-4-pyridinecarboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₄ClNO₄S
MW: 303.7619 g/mol
MDL Number: MFCD00723388
CAS Number: 314744-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR