3-Amino-4-pyrrolidin-1-ylbenzoic acid

Supplier: Matrix Scientific

032987-500MG
101834-800EA 230 USD
101834-800
3-Amino-4-pyrrolidin-1-ylbenzoic acid
3-Amino-4-pyrrolidin-1-ylbenzoic acid

Hazard: Irritant

Formula: C₁₁H₁₄N₂O₂
MDL Number: MFCD06010205
CAS Number: 123986-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR