2-Isopropyl-2H-pyrazole-3-carbaldehyde

Supplier: Matrix Scientific

Synonyms: 1-Isopropyl-1H-pyrazole-5-carbaldehyde

032980-500MG
101834-786EA 282.9 USD
101834-786
2-Isopropyl-2H-pyrazole-3-carbaldehyde
2-Isopropyl-2H-pyrazole-3-carbaldehyde

Hazard: Irritant

Formula: C₇H₁₀N₂O
MW: 138.17 g/mol
MDL Number: MFCD03130042
CAS Number: 100305-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR