3-(4-Methyl-4H-[1,2,4]triazol-3-yl)phenylamine

Supplier: Matrix Scientific

Synonyms: 3-(4-Methyl-4H-1,2,4-triazol-3-yl)aniline

032976-500MG
101834-778EA 230 USD
101834-778
3-(4-Methyl-4H-[1,2,4]triazol-3-yl)phenylamine
3-(4-Methyl-4H-[1,2,4]triazol-3-yl)phenylamine

Hazard: Irritant

Formula: C₉H₁₀N₄
MW: 174.21 g/mol
MDL Number: MFCD02853082
CAS Number: 252928-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR