1-(4-Chloromethylphenyl)pyrrolidin-2-one

Supplier: Matrix Scientific

032962-500MG
101834-750EA 188.6 USD
101834-750
1-(4-Chloromethylphenyl)pyrrolidin-2-one
1-(4-Chloromethylphenyl)pyrrolidin-2-one

Hazard: Irritant

Formula: C₁₁H₁₂ClNO
MDL Number: MFCD03196493
CAS Number: 36152-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR