3-{[(Benzo[1,3]dioxol-5-ylmethyl)amino]methyl}-7-methyl-1H-quinolin-2-one

Supplier: Matrix Scientific

032911-500MG
101834-656EA 230 USD
101834-656
3-{[(Benzo[1,3]dioxol-5-ylmethyl)amino]methyl}-7-methyl-1H-quinolin-2-one
3-{[(Benzo[1,3]dioxol-5-ylmethyl)amino]methyl}-7-methyl-1H-quinolin-2-one

Hazard: Irritant

Formula: C₁₉H₁₈N₂O₃
MDL Number: MFCD02748604
CAS Number: 484025-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR