5-(1H-Indol-3-ylmethyl)[1,3,4]thiadiazol-2-ylamine

Supplier: Matrix Scientific

032767-500MG
101834-392EA 230 USD
101834-392
5-(1H-Indol-3-ylmethyl)[1,3,4]thiadiazol-2-ylamine
5-(1H-Indol-3-ylmethyl)[1,3,4]thiadiazol-2-ylamine

Hazard: Irritant

Formula: C₁₁H₁₀N₄S
MDL Number: MFCD01430063
CAS Number: 153595-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR