3-(4-Methoxy-2-methylphenyl)propionic acid

Supplier: Matrix Scientific

032735-500MG
101834-332EA 282.9 USD
101834-332
3-(4-Methoxy-2-methylphenyl)propionic acid
3-(4-Methoxy-2-methylphenyl)propionic acid

Hazard: Irritant

Formula: C₁₁H₁₄O₃
Melting Pt: 98…99 °C
MDL Number: MFCD00795968
CAS Number: 28353-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR