4-(2-1H-Indol-3-ylacetylamino)butyric acid

Supplier: Matrix Scientific

032700-500MG
101834-268EA 184 USD
101834-268
4-(2-1H-Indol-3-ylacetylamino)butyric acid
4-(2-1H-Indol-3-ylacetylamino)butyric acid

Hazard: Irritant

Formula: C₁₄H₁₆N₂O₃
MDL Number: MFCD01593582
CAS Number: 79438-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR