1-(2-Bromoethoxy)-3-methoxybenzene

Supplier: Matrix Scientific

Synonyms: 3-(2-Bromoethoxy)anisole

032661-500MG
101834-198EA 307.69 USD
101834-198
1-(2-Bromoethoxy)-3-methoxybenzene
1-(2-Bromoethoxy)-3-methoxybenzene

Hazard: Irritant

Formula: C₉H₁₁O₂Br
MW: 231.0864 g/mol
MDL Number: MFCD00980056
CAS Number: 3245-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR