3-(4-Methylphenyl)-1H-pyrazol-5-amine

Supplier: Matrix Scientific

Synonyms: 5-Amino-3-(4-methylphenyl)pyrazole

032657-500MG
101834-190EA 20.7 USD
101834-190
3-(4-Methylphenyl)-1H-pyrazol-5-amine
3-(4-Methylphenyl)-1H-pyrazol-5-amine

Hazard: Irritant

Formula: C₁₀H₁₁N₃
MDL Number: MFCD00084887
CAS Number: 78597-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR