1-(2-Aminoethyl)cyclohexanol

Supplier: Matrix Scientific

032637-500MG
101834-150EA 438.46 USD
101834-150
1-(2-Aminoethyl)cyclohexanol
1-(2-Aminoethyl)cyclohexanol

Hazard: Irritant

Formula: C₈H₁₇NO
MDL Number: MFCD00778619
CAS Number: 39884-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR