2-Amino-4-(2-pyridyl)thiazole

Supplier: Matrix Scientific

Synonyms: 4-(Pyridin-2-yl)thiazol-2-amine

032629-500MG
101834-132EA 407.69 USD
101834-132
2-Amino-4-(2-pyridyl)thiazole
2-Amino-4-(2-pyridyl)thiazole

Hazard: Irritant

Formula: C₈H₇N₃S
MW: 177.22 g/mol
MDL Number: MFCD00460417
CAS Number: 30235-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR