2-Piperidin-1-ylmethylmorpholine

Supplier: Matrix Scientific

032626-500MG
101834-130EA 307.69 USD
101834-130
2-Piperidin-1-ylmethylmorpholine
2-Piperidin-1-ylmethylmorpholine

Hazard: Irritant

Formula: C₁₀H₂₀N₂O
MDL Number: MFCD00770012
CAS Number: 122894-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR