4-(2-Benzooxazol-2-ylethyl)phenylamine

Supplier: Matrix Scientific

032621-500MG
101834-120EA 307.69 USD
101834-120
4-(2-Benzooxazol-2-ylethyl)phenylamine
4-(2-Benzooxazol-2-ylethyl)phenylamine

Hazard: Irritant

Formula: C₁₅H₁₄N₂O
MDL Number: MFCD00608319
CAS Number: 39921-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR