2,2,5,5-Tetramethyl-2,5-dihydro-1H-pyrrole-3-carboxylic acid

Supplier: Matrix Scientific

032620-500MG
101834-118EA 230 USD
101834-118
2,2,5,5-Tetramethyl-2,5-dihydro-1H-pyrrole-3-carboxylic acid
2,2,5,5-Tetramethyl-2,5-dihydro-1H-pyrrole-3-carboxylic acid

Hazard: Irritant

Formula: C₉H₁₅NO₂
MDL Number: MFCD00568208
CAS Number: 76194-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR