1-Isobutyl-2,3,4,9-tetrahydro-1H-β-carboline-3-carboxylic acid acetate

Supplier: Matrix Scientific

032597-500MG
101834-076EA 230 USD
101834-076
1-Isobutyl-2,3,4,9-tetrahydro-1H-β-carboline-3-carboxylic acid acetate
1-Isobutyl-2,3,4,9-tetrahydro-1H-beta-carboline-3-carboxylic acid acetate

Hazard: Irritant

Formula: C₁₈H₂₄N₂O₄
MDL Number: MFCD11506410
CAS Number: 1139669-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR