3-Chloro-2,6-difluorobenzotrifluoride ≥95%

Supplier: Matrix Scientific

032577-1G
101834-032EA 169.05 USD
101834-032
3-Chloro-2,6-difluorobenzotrifluoride ≥95%
3-Chloro-2,6-difluorobenzotrifluoride

Hazard: Irritant

Formula: C₇H₂ClF₅
MDL Number: MFCD11226546
CAS Number: 1099597-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR