1,3-Difluoro-2-(2,2,2-trifluoroethyl)benzene ≥95%

Supplier: Matrix Scientific

032572-1G
101834-022EA 250.77 USD
101834-022
1,3-Difluoro-2-(2,2,2-trifluoroethyl)benzene ≥95%
1,3-Difluoro-2-(2,2,2-trifluoroethyl)benzene

Hazard: Irritant

Formula: C₈H₅F₅
MDL Number: MFCD11226576
CAS Number: 1099597-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR