1-Bromo-4-(3,3,3-trifluoropropyl)benzene ≥95%

Supplier: Matrix Scientific

032570-1G
101834-018EA 80.5 USD
101834-018
1-Bromo-4-(3,3,3-trifluoropropyl)benzene ≥95%
1-Bromo-4-(3,3,3-trifluoropropyl)benzene

Hazard: Irritant

Formula: C₉H₈BrF₃
MDL Number: MFCD11226575
CAS Number: 1099597-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR