1-Chloro-3-fluoro-2-(2,2,2-trifluoroethyl)benzene ≥95%
Supplier: Matrix Scientific
032568-1G
101834-014EA
235.38
USD
101834-014
1-Chloro-3-fluoro-2-(2,2,2-trifluoroethyl)benzene ≥95%
1-Chloro-3-fluoro-2-(2,2,2-trifluoroethyl)benzene
Hazard: Irritant
Formula:
C₈H₅ClF₄ |
MDL Number:
MFCD11226574 CAS Number: 1099597-29-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...