5-(5-Ethylthien-3-yl)-4-methyl-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030301-500MG
101829-864EA 193.85 USD
101829-864
5-(5-Ethylthien-3-yl)-4-methyl-4H-1,2,4-triazole-3-thiol
5-(5-Ethylthien-3-yl)-4-methyl-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₉H₁₁N₃S₂
MDL Number: MFCD03423419
CAS Number: 588674-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR