4-Isopropyl-5-(5-methylthien-3-yl)-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030299-500MG
101829-860EA 193.85 USD
101829-860
4-Isopropyl-5-(5-methylthien-3-yl)-4H-1,2,4-triazole-3-thiol
4-Isopropyl-5-(5-methylthien-3-yl)-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₀H₁₃N₃S₂
MDL Number: MFCD03423418
CAS Number: 588682-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR